Assay Details
Gene Symbol: TRMT10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498449.4 | XP_001498499.1 | 356 | 74 | ||
XM_005608596.3 | XP_005608653.1 | 1629 | 74 | |||
XM_005608597.3 | XP_005608654.1 | 1058 | 74 | |||
XM_005608599.3 | XP_005608656.1 | 1283 | 74 | |||
XM_005608600.3 | XP_005608657.1 | 952 | 74 | |||
XM_005608601.3 | XP_005608658.1 | 1629 | 74 | |||
XM_014738746.2 | XP_014594232.1 | 261 | 74 | |||
XM_023637662.1 | XP_023493430.1 | 1034 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
