Assay Details
Gene Symbol: EIF4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637674.1 | XP_023493442.1 | 653 | 84 | ||
XM_023637675.1 | XP_023493443.1 | 1002 | 84 | |||
GenBank mRNA | JL616474.1 | - | 6 - 7 | 564 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
