Assay Details
Gene Symbol: SNCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608622.3 | XP_005608679.1 | 344 | 63 | ||
XM_014738855.2 | XP_014594341.1 | 335 | 63 | |||
XM_014738856.2 | XP_014594342.1 | 550 | 63 | |||
XM_014738857.2 | XP_014594343.1 | 303 | 63 | |||
XM_014738858.2 | XP_014594344.1 | 321 | 63 | |||
XM_014738860.2 | XP_014594346.1 | 312 | 63 | |||
XM_023637715.1 | XP_023493483.1 | 306 | 63 | |||
XM_023637716.1 | XP_023493484.1 | 303 | 63 | |||
XM_023637717.1 | XP_023493485.1 | 292 | 63 | |||
XM_023637718.1 | XP_023493486.1 | 303 | 63 | |||
XM_023637719.1 | XP_023493487.1 | 345 | 63 | |||
XM_023637720.1 | XP_023493488.1 | 309 | 63 | |||
XM_023637721.1 | XP_023493489.1 | 219 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
