Assay Details
Gene Symbol: NUDT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608646.3 | XP_005608703.2 | 886 | 69 | ||
XM_005608648.3 | XP_005608705.2 | 656 | 69 | |||
XM_023637753.1 | XP_023493521.1 | 892 | 69 | |||
GenBank mRNA | JL616066.1 | - | 5 - 6 | 588 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
