Assay Details
Gene Symbol: GPAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608660.3 | XP_005608717.1 | 1655 | 66 | ||
XM_005608661.3 | XP_005608718.1 | 1503 | 66 | |||
XM_023637800.1 | XP_023493568.1 | 1805 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
