Assay Details
Gene Symbol: ABRAXAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637801.1 | XP_023493569.1 | 547 | 67 | ||
XM_023637802.1 | XP_023493570.1 | 545 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
