Assay Details
Gene Symbol: SEPTIN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491198.5 | XP_001491248.1 | 834 | 59 | ||
XM_005608688.3 | XP_005608745.1 | 835 | 59 | |||
XM_005608689.3 | XP_005608746.1 | 835 | 59 | |||
XM_005608690.3 | XP_005608747.1 | 834 | 59 | |||
XM_005608691.3 | XP_005608748.1 | 834 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
