Assay Details
Gene Symbol: COX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488484.5 | XP_001488534.3 | 848 | 102 | ||
XM_005608725.3 | XP_005608782.2 | 850 | 102 | |||
XM_014738806.2 | XP_014594292.1 | 534 | 102 | |||
GenBank mRNA | JL622018.1 | - | 4 - 5 | 517 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
