Assay Details
Gene Symbol: TMPRSS11F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638028.1 | XP_023493796.1 | 912 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
