Assay Details
Gene Symbol: NOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916805.5 | XP_001916840.4 | 2163 | 60 | ||
GenBank mRNA | JL625024.1 | - | 4 - 5 | 758 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
