Assay Details
Gene Symbol: COX7B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608792.3 | XP_005608849.1 | 118 | 82 | ||
XM_005608793.3 | XP_005608850.1 | 118 | 82 | |||
XM_014738586.2 | XP_014594072.1 | 118 | 82 | |||
XM_023638143.1 | XP_023493911.1 | 118 | 82 | |||
XM_023638144.1 | XP_023493912.1 | 118 | 82 | |||
XM_023638145.1 | XP_023493913.1 | 118 | 82 | |||
XM_023638146.1 | XP_023493914.1 | 118 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005608792.3, XM_005608793.3, XM_014738586.2, XM_023638143.1, XM_023638144.1, XM_023638145.1, XM_023638146.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
