Assay Details
Gene Symbol: GRXCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496545.5 | XP_001496595.3 | 891 | 81 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
