Assay Details
Gene Symbol: KLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014738712.2 | XP_014594198.1 | 1200 | 63 | ||
XM_023638230.1 | XP_023493998.1 | 1166 | 63 | |||
XM_023638231.1 | XP_023493999.1 | 1086 | 63 | |||
XM_023638232.1 | XP_023494000.1 | 919 | 63 | |||
XM_023638233.1 | XP_023494001.1 | 1127 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
