Assay Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608836.3 | XP_005608893.1 | 529 | 74 | ||
XM_014738725.2 | XP_014594211.1 | 768 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
