Assay Details
Gene Symbol: PCDH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638257.1 | XP_023494025.1 | 4886 | 70 | ||
XM_023638259.1 | XP_023494027.1 | 4862 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
