Assay Details
Gene Symbol: QDPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638310.1 | XP_023494078.1 | 248 | 93 | ||
XM_023638311.1 | XP_023494079.1 | 249 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
