Assay Details
Gene Symbol: CYTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499643.4 | XP_001499693.4 | 172 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
