Assay Details
Gene Symbol: ZBTB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492205.4 | XP_001492255.1 | 1898 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001492205.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
