Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023632770.1 | XP_023488538.1 | 1560 | 83 | ||
XM_023632771.1 | XP_023488539.1 | 1557 | 83 | |||
XM_023632772.1 | XP_023488540.1 | 1550 | 83 | |||
XM_023632773.1 | XP_023488541.1 | 1547 | 83 | |||
XM_023632774.1 | XP_023488542.1 | 1562 | 83 | |||
XM_023632775.1 | XP_023488543.1 | 1554 | 83 | |||
XM_023632776.1 | XP_023488544.1 | 1558 | 83 | |||
XM_023632777.1 | XP_023488545.1 | 1540 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
