Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023632770.1 | XP_023488538.1 | 1816 | 70 | ||
XM_023632771.1 | XP_023488539.1 | 1813 | 70 | |||
XM_023632772.1 | XP_023488540.1 | 1806 | 70 | |||
XM_023632773.1 | XP_023488541.1 | 1803 | 70 | |||
XM_023632774.1 | XP_023488542.1 | 1818 | 70 | |||
XM_023632777.1 | XP_023488545.1 | 1796 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
