Assay Details
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607979.3 | XP_005608036.1 | 589 | 71 | ||
XM_005607980.3 | XP_005608037.1 | 587 | 71 | |||
XM_005607981.3 | XP_005608038.1 | 588 | 71 |
