Assay Details
Gene Symbol: UBE2T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494968.4 | XP_001495018.2 | 622 | 68 | ||
XM_023632657.1 | XP_023488425.1 | 621 | 68 | |||
XM_023632658.1 | XP_023488426.1 | 547 | 68 | |||
XM_023632659.1 | XP_023488427.1 | 620 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
