Assay Details
Gene Symbol: MPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023633142.1 | XP_023488910.1 | 445 | 81 | ||
GenBank mRNA | JL624900.1 | - | 1 - 2 | 153 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
