Assay Details
Gene Symbol: CCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489474.6 | XP_001489524.3 | 338 | 76 | ||
XM_005608132.3 | XP_005608189.1 | 443 | 76 | |||
XM_023632933.1 | XP_023488701.1 | 298 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001489474.6, XM_005608132.3, XM_023632933.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
