Assay Details
Gene Symbol: AKAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608180.3 | XP_005608237.2 | 5250 | 86 | ||
XM_005608181.3 | XP_005608238.2 | 5220 | 86 | |||
XM_023633109.1 | XP_023488877.1 | 5384 | 86 | |||
XM_023633110.1 | XP_023488878.1 | 5254 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
