Assay Details
Gene Symbol: EPM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023633123.1 | XP_023488891.1 | 492 | 96 | ||
XM_023633124.1 | XP_023488892.1 | 731 | 96 | |||
XM_023633125.1 | XP_023488893.1 | 492 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
