Assay Details
Gene Symbol: EPM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023633123.1 | XP_023488891.1 | 729 | 127 | ||
XM_023633124.1 | XP_023488892.1 | 968 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() |
