Assay Details
Gene Symbol: NAPEPLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488764.4 | XP_001488814.1 | 496 | 69 | ||
XM_005609045.3 | XP_005609102.1 | 472 | 69 | |||
XM_005609046.3 | XP_005609103.1 | 602 | 69 | |||
XM_014738935.2 | XP_014594421.1 | 2279 | 69 | |||
XM_023638843.1 | XP_023494611.1 | 421 | 69 | |||
XM_023638844.1 | XP_023494612.1 | 332 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
