Assay Details
Gene Symbol: CBLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491503.6 | XP_001491553.1 | 407 | 99 | ||
XM_005609075.3 | XP_005609132.1 | 404 | 99 | |||
XM_023638898.1 | XP_023494666.1 | 909 | 99 | |||
XM_023638899.1 | XP_023494667.1 | 906 | 99 | |||
XM_023638900.1 | XP_023494668.1 | 384 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
