Assay Details
Gene Symbol: UBE2D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638978.1 | XP_023494746.1 | 523 | 69 | ||
XM_023638979.1 | XP_023494747.1 | 308 | 69 | |||
XM_023638980.1 | XP_023494748.1 | 523 | 69 | |||
GenBank mRNA | JL627710.1 | - | 1 - 2 | 597 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
