Assay Details
Gene Symbol: SLC25A41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495697.5 | XP_001495747.3 | 1689 | 75 | ||
XM_023644579.1 | XP_023500347.1 | 1690 | 75 | |||
XM_023644580.1 | XP_023500348.1 | 1691 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
