Assay Details
Gene Symbol: CRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916887.5 | XP_001916922.2 | 191 | 72 | ||
XM_023644582.1 | XP_023500350.1 | 191 | 72 | |||
XM_023644583.1 | XP_023500351.1 | 301 | 72 | |||
XM_023644584.1 | XP_023500352.1 | 276 | 72 | |||
XM_023644585.1 | XP_023500353.1 | 285 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001916887.5, XM_023644582.1, XM_023644583.1, XM_023644584.1, XM_023644585.1 transcripts.
Set Membership: |
![]() ![]() |
