Assay Details
Gene Symbol: CRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916887.5 | XP_001916922.2 | 902 | 77 | ||
XM_023644582.1 | XP_023500350.1 | 918 | 77 | |||
XM_023644583.1 | XP_023500351.1 | 1012 | 77 | |||
XM_023644584.1 | XP_023500352.1 | 987 | 77 | |||
XM_023644585.1 | XP_023500353.1 | 996 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
