Assay Details
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498350.6 | XP_001498400.4 | 2090 | 70 | ||
XM_014741245.2 | XP_014596731.2 | 2092 | 70 | |||
XM_023644668.1 | XP_023500436.1 | 2217 | 70 | |||
GenBank mRNA | JL621991.1 | - | 4 - 5 | 1145 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
