Assay Details
Gene Symbol: CCDC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005611530.3 | XP_005611587.2 | 1964 | 81 | ||
XM_023646435.1 | XP_023502203.1 | 1864 | 81 | |||
XM_023646436.1 | XP_023502204.1 | 1730 | 81 | |||
XM_023646437.1 | XP_023502205.1 | 1692 | 81 | |||
XM_023646438.1 | XP_023502206.1 | 1861 | 81 | |||
XM_023646439.1 | XP_023502207.1 | 1801 | 81 | |||
XM_023646440.1 | XP_023502208.1 | 1780 | 81 | |||
XM_023646441.1 | XP_023502209.1 | 1668 | 81 | |||
XM_023646442.1 | XP_023502210.1 | 1558 | 81 | |||
XM_023646443.1 | XP_023502211.1 | 1727 | 81 | |||
XM_023646444.1 | XP_023502212.1 | 1596 | 81 | |||
XM_023646445.1 | XP_023502213.1 | 1534 | 81 | |||
XM_023646446.1 | XP_023502214.1 | 1646 | 81 | |||
XM_023646447.1 | XP_023502215.1 | 1629 | 81 | |||
XM_023646448.1 | XP_023502216.1 | 1508 | 81 | |||
XM_023646449.1 | XP_023502217.1 | 1864 | 81 | |||
GenBank mRNA | HQ890038.1 | - | 5 - 6 | 746 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
