Assay Details
Gene Symbol: BIRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499875.5 | XP_001499925.1 | 3779 | 80 | ||
XM_023644710.1 | XP_023500478.1 | 3763 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
