Assay Details
Gene Symbol: BIRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023644711.1 | XP_023500479.1 | 2731 | 73 | ||
GenBank mRNA | HQ890264.1 | - | 2 - 3 | 96 | 73 | |
JL616441.1 | - | 4 - 5 | 1055 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
