Assay Details
Gene Symbol: AASDHPPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005611544.3 | XP_005611601.1 | 871 | 72 | ||
GenBank mRNA | JL624010.1 | - | 4 - 5 | 483 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
