Assay Details
Gene Symbol: NXPE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005611582.3 | XP_005611639.1 | 1381 | 74 | ||
XM_005611586.3 | XP_005611643.1 | 1381 | 74 | |||
XM_005611587.3 | XP_005611644.1 | 269 | 74 | |||
XM_014741370.2 | XP_014596856.1 | 1369 | 74 | |||
XM_014741371.2 | XP_014596857.1 | 1307 | 74 | |||
XM_023644853.1 | XP_023500621.1 | 1150 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
