Assay Details
Gene Symbol: TBRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001505067.5 | XP_001505117.1 | 970 | 121 | ||
XM_023645057.1 | XP_023500825.1 | 896 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
