Assay Details
Gene Symbol: MSANTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917864.5 | XP_001917899.3 | 645 | 105 | ||
XM_023645061.1 | XP_023500829.1 | 801 | 105 | |||
XM_023645063.1 | XP_023500831.1 | 4325 | 105 | |||
XM_023645064.1 | XP_023500832.1 | 520 | 105 | |||
XM_023645065.1 | XP_023500833.1 | 460 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
