Assay Details
Gene Symbol: FOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502385.5 | XP_001502435.2 | 1563 | 63 | ||
XM_005611694.3 | XP_005611751.1 | 1575 | 63 | |||
XM_014741313.2 | XP_014596799.1 | 1464 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
