Assay Details
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014741275.2 | XP_014596761.1 | 1754 | 77 | ||
XM_023645110.1 | XP_023500878.1 | 3811 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
