Assay Details
Gene Symbol: DCPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001505098.4 | XP_001505148.1 | 231 | 78 | ||
XM_005611695.3 | XP_005611752.1 | 233 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
