Assay Details
Gene Symbol: ADGRE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005611833.3 | XP_005611890.1 | 772 | 68 | ||
XM_023645192.1 | XP_023500960.1 | 787 | 68 | |||
XM_023645193.1 | XP_023500961.1 | 1003 | 68 | |||
XM_023645194.1 | XP_023500962.1 | 630 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
