Assay Details
Gene Symbol: CLEC17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495641.6 | XP_001495691.3 | 486 | 73 | ||
XM_005611836.3 | XP_005611893.1 | 408 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
