Assay Details
Gene Symbol: TECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498371.6 | XP_001498421.1 | 257 | 111 | ||
XM_005611839.3 | XP_005611896.1 | 196 | 111 | |||
XM_014741446.2 | XP_014596932.1 | 309 | 111 | |||
XM_023645199.1 | XP_023500967.1 | 178 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
