Assay Details
Gene Symbol: PRKACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001914777.5 | XP_001914812.1 | 1042 | 50 | ||
XM_005611840.3 | XP_005611897.1 | 2602 | 50 | |||
XM_023645231.1 | XP_023500999.1 | 655 | 50 |

More Information
Set Membership: |
![]() ![]() ![]() |
