Assay Details
Gene Symbol: STX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504875.5 | XP_001504925.3 | 414 | 111 | ||
XM_023645278.1 | XP_023501046.1 | 414 | 111 | |||
XM_023645279.1 | XP_023501047.1 | 414 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
