Assay Details
Gene Symbol: PRDX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645315.1 | XP_023501083.1 | 527 | 63 | ||
XM_023645316.1 | XP_023501084.1 | 527 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
